missing translation for 'onlineSavingsMsg'
Learn More

Arachidic Acid Methyl Ester, TRC

Product Code. 30135986
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135986 25 g 25g
30012391 1 g 1g
17968864 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135986 Supplier Toronto Research Chemicals Supplier No. TRCA76498025G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-28-1
Molecular Formula C21 H42 O2
Molecular Weight (g/mol) 326.56
Synonym Eicosanoic Acid Methyl Ester, Eicosanoic acid, methyl ester, Arachic acid methyl ester, Arachidic acid methyl ester, C20:0 FAME, Methyl arachate, Methyl arachidate, Methyl eicosanate, Methyl eicosanoate, n-Eicosanoic acid methyl ester
IUPAC Name methyl icosanoate
SMILES CCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C21H42O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h3-20H2,1-2H3
Formula Weight 326.3185
Percent Purity >95%
Chemical Name or Material Arachidic Acid Methylester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.