missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This antibody has been tested in direct-ELISA.
This maternal effect gene is oocyte-specific and encodes a protein that is thought to function in the initiation of embryogenesis. A similar protein in mouse is required for female fertility. [provided by RefSeq, Jul 2013].
Specifications
Specifications
| Antigen | Arabidopsis thaliana ZAR1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 2.66 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene Alias | oocyte-specific maternal effect factor; Z3CXXC6; ZAR1; zinc finger, 3CxxC-type 6; zygote arrest 1; zygote arrest protein 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant Arabidopsis thaliana Receptor protein kinase-like protein ZAR1 protein (167-278aa). |
| Purification Method | Protein G |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
