missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARA54 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33230-100ul
This item is not returnable.
View return policy
Description
ARA54 Monoclonal antibody specifically detects ARA54 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ARA54 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Androgen receptor-associated protein 54, ARA54androgen receptor associated protein 54, EC 6.3.2.-, HFB30E3 ubiquitin-protein ligase RNF14, ring finger protein 14FLJ26004, TRIAD2, Triad2 protein | |
| A synthetic peptide corresponding to a sequence within amino acids 300-400 of human ARA54 (Q9UBS8).,, Sequence:, LDLMADVVYCPRPCCQLPVMQEPGCTMGICSSCNFAFCTLCRLTYHGVSPCKVTAEKLMDLRNEYLQADEANKRLLDQRYGKRVIQKALEEMESKEWLEKN | |
| 100 μL | |
| Signal Transduction, Zinc Finger | |
| 9604 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction