missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AR Mouse anti-Human, Clone: AR441 + DHTR/882, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14567.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against human AR.
- High specificity
- High reactivity
Specifications
| AR | |
| Monoclonal | |
| Unconjugated | |
| Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| P10275 | |
| AR | |
| A synthetic peptide corresponding to amino acids 302-318 of human AR (AR441) and recombinant protein corresponding to full length human AR (DHTR/882). | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| AR441 + DHTR/882 | |
| Mouse monoclonal antibody raised against human AR. | |
| In 1 mM PBS (0.05% BSA, 0.05% sodium azide). | |
| AIS/DHTR/HUMARA/KD/NR3C4/SBMA/SMAX1/TFM | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 367 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction