missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AR-C155858, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | AR-C155858 |
| CAS | 496791-37-8 |
| Molecular Formula | C21 H27 N5 O5 S |
| Quantity | 10 mg |
| Synonym | (S)-6-[(3,5-Dimethyl-1H-pyrazol-4-yl)methyl]-5-[(4-hydroxyisoxazolidin-2-yl)carbonyl]-1-isobutyl-3-methylthieno[2,3-d]pyrimidine-2,4(1H,3H)-dione, 2-[[6-[(3,5-dimethyl-1H-pyrazol-4-yl)methyl]-1,2,3,4-tetrahydro-3-methyl-1-(2-methylpropyl)-2,4-dioxothieno[2,3-d]pyrimidin-5-yl]carbonyl]-4-isoxazolidinol, (S)-6-[(3,5-Dimethyl-1H-pyrazol-4-yl)methyl]-5-[(4-hydroxyisoxazolidin-2-yl)carbonyl]-1-isobutyl-3-methylthieno[2,3-d]pyrimidine-2,4(1H,3H)-dione, AR-C 155858 |
| InChI Formula | InChI=1S/C21H27N5O5S/c1-10(2)7-25-20-17(18(28)24(5)21(25)30)16(19(29)26-8-13(27)9-31-26)15(32-20)6-14-11(3)22-23-12(14)4/h10,13,27H,6-9H2,1-5H3,(H,22,23)/t13-/m0/s1 |
| SMILES | CC(C)CN1C(=O)N(C)C(=O)c2c1sc(Cc3c(C)n[nH]c3C)c2C(=O)N4C[C@H](O)CO4 |
| IUPAC Name | 6-[(3,5-dimethyl-1H-pyrazol-4-yl)methyl]-5-[(4S)-4-hydroxy-1,2-oxazolidine-2-carbonyl]-3-methyl-1-(2-methylpropyl)thieno[2,3-d]pyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 461.535 |
| Formula Weight | 461.173 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction