missing translation for 'onlineSavingsMsg'
Learn More

AR-C155858, MedChemExpress

Product Code. 16471329
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16471329

Brand: Medchem Express HY13248

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AR-C155858 is a selective monocarboxylate transporter MCT1 and MCT2 inhibitor with Kis of 2.3 nM and 10 nM, respectively.

AR-C155858 is a selective monocarboxylate transporter MCT1 and MCT2 inhibitor with Kis of 2.3 nM and 10 nM, respectively.

Specifications

Chemical Name or Material AR-C155858
Physical Form Powder
CAS 496791-37-8
Color Off-White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C21H27N5O5S
Solubility Information DMSO : ≥ 50 mg/mL (108.34 mM)
SMILES O=C(C1=C2SC(CC3=C(NN=C3C)C)=C1C(N4C[C@@H](CO4)O)=O)N(C(N2CC(C)C)=O)C
Molecular Weight (g/mol) 461.53
Formula Weight 461.53
Percent Purity 95.56%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.