missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AR-C 118925XX
GREENER_CHOICE

Product Code. 18716576
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 18716576

Brand: Tocris Bioscience™ 4890/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective and competitive P2Y2 antagonist

AR-C 118925XX is a potent, selective, competitive and reversible P2Y2 receptor antagonist (pA2 = 8.43). Inactive against a panel of 37 other receptors at a concentration of 10 μM. AR-C 118925XX inhibits ATP-γS-induced mucin secretion in bronchial epithelial cells (IC50 = 1 μM). Inhibits P2Y2 receptor-induced β-arrestin translocation in vitro (pA2 = 37.2-51.3 nM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[[5-(2,8-Dimethyl-5H-dibenzo[a,d]cyclohepten-5-yl)-3,4-dihydro-2-oxo-4-thioxo-1(2H)-pyrimidinyl]methyl]-N-2H-tetrazol-5-yl-2-furancarboxamide
CAS 216657-60-2
Quantity 5 mg
Target P2Y Receptor Antagonists
Molecular Formula C28H23N7O3S
Formula Weight 537.59
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.