missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AR 231453, MedChemExpress
Description
AR 231453 is a potent, specific and orally available GPR119 agonist. AR 231453 can stimulate β-cell replication and improve islet graft function s.
Specifications
Specifications
| Chemical Name or Material | AR 231453 |
| CAS | 733750-99-7 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H24FN7O5S |
| Quantity | 5 mg |
| Solubility Information | DMSO : 25 mg/mL (49.45 mM; Need ultrasonic) |
| SMILES | CC(C)C1=NOC(C2CCN(C3=NC=NC(NC4=CC=C(S(=O)(C)=O)C=C4F)=C3[N+]([O-])=O)CC2)=N1 |
| Molecular Weight (g/mol) | 505.52 |
| Formula Weight | 505.52 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?