missing translation for 'onlineSavingsMsg'
Learn More

AR-13324 M1 metabolite, MedChemExpress

Product Code. 16471547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471547 50 mg 50mg
16455358 10 mg 10mg
16428077 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16471547 Supplier Medchem Express Supplier No. HY12798C
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AR-13324 M1 metabolite is a hydrolysis metabolite of AR-13324 mesylate. AR-13324 is a inhibitor of ROCK kinase.

AR-13324 M1 metabolite is a hydrolysis metabolite of AR-13324 mesylate. AR-13324 is a inhibitor of ROCK kinase.

Specifications

Chemical Name or Material AR-13324 M1 metabolite
CAS 2309668-15-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H19N3O2
Quantity 50 mg
Solubility Information DMSO : ≥ 41.83 mg/mL (130.16 mM)
SMILES OCC1=CC=C([C@@H](CN)C(NC2=CC(C=CN=C3)=C3C=C2)=O)C=C1
Molecular Weight (g/mol) 321.37
Formula Weight 321.37
Percent Purity 95.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.