missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aquaporin 8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576415
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Aquaporin-8 is a protein that in humans is encoded by the AQP8 gene. Aquaporin 8 (AQP8) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 8 mRNA is found in pancreas and colon but not other tissues. Forms a water-specific channel; mercury-sensitive. Not permeable to glycerol or urea. Expressed only in pancreas and colon.
Specifications
| Aquaporin 8 | |
| Polyclonal | |
| Unconjugated | |
| Aqp8 | |
| AI255744; AQP8; AQP-8; aquaporin 8; Aquaporin8; aquaporin-8; membrane water channel; MGC93492; water channel | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11833, 29172, 343 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94778, P56404, P56405 | |
| Aqp8 | |
| Synthetic peptide of human AQP8. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction