missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aquaporin 8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578817
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human A431 whole cell, human PC-3 whole cell.
Aquaporin-8 is a protein that in humans is encoded by the AQP8 gene. Aquaporin 8 (AQP8) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 8 mRNA is found in pancreas and colon but not other tissues. Forms a water-specific channel; mercury-sensitive. Not permeable to glycerol or urea. Expressed only in pancreas and colon.
Specifications
| Aquaporin 8 | |
| Polyclonal | |
| Unconjugated | |
| Aqp8 | |
| AI255744; AQP8; AQP-8; aquaporin 8; Aquaporin8; aquaporin-8; membrane water channel; MGC93492; water channel | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11833, 29172, 343 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| O94778, P56404, P56405 | |
| Aqp8 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human Aquaporin 8 (1-19aa MSGEIAMCEPEFGNDKARE). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction