missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aquaporin 8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597109
This item is not returnable.
View return policy
Description
Positive Samples: Mouse liver, Mouse heart, Rat liver; Cellular Location: Membrane, Multi-pass membrane protein Immunogen sequence: MLIGGLNLVM LLPYWVSQLL GGMLGAALAK AVSPEERFWN ASGAAFVTVQ EQGQVAGALV AEIILTTLLA LAVCMGAINE KTKGPLAPFS IGFAVTVDIL A.
Aquaporin-8 is a protein that in humans is encoded by the AQP8 gene. Aquaporin 8 (AQP8) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 8 mRNA is found in pancreas and colon but not other tissues. Forms a water-specific channel; mercury-sensitive. Not permeable to glycerol or urea. Expressed only in pancreas and colon.
Specifications
| Aquaporin 8 | |
| Polyclonal | |
| Unconjugated | |
| Aqp8 | |
| AI255744; AQP8; AQP-8; aquaporin 8; Aquaporin8; aquaporin-8; membrane water channel; MGC93492; water channel | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11833, 29172, 343 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.09 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O94778, P56404, P56405 | |
| Aqp8 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction