missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-8 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92557-0.1ml
This item is not returnable.
View return policy
Description
Aquaporin-8 Polyclonal antibody specifically detects Aquaporin-8 in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Aquaporin-8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| AQP-8, aquaporin 8, aquaporin-8 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Aquaporin-8 (NP_001160.2). MLIGGLNLVMLLPYWVSQLLGGMLGAALAKAVSPEERFWNASGAAFVTVQEQGQVAGALVAEIILTTLLALAVCMGAINEKTKGPLAPFSIGFAVTVDILA | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 343 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction