missing translation for 'onlineSavingsMsg'
Learn More

Aquaporin-8 Antibody (1A1), Novus Biologicals™

Product Code. 18372058 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18372058 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18372058 Supplier Novus Biologicals Supplier No. H00000343M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aquaporin-8 Monoclonal antibody specifically detects Aquaporin-8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin-8
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 1A1
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH40630
Gene Alias AQP-8, aquaporin 8, aquaporin-8
Host Species Mouse
Immunogen AQP8 (AAH40630, 1 a.a. ~ 255 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MCEPEFGNDKAREPSVGGRWRVSWYERFVQPCLVELLGSALFIFIGCLSVIENGTDTGLLQPALAHGLALGLVIATLGNISGGHFNPAVSLAAMLIGGLNLVMLLPYWVSQLLGGMLGAALAKAVSPEERFWNASGAAFVTVQEQGQVAGALVAEIILTTLLALAVCMGAINEKTKGPLAPFSIGFAVTVDILAGGPVSGGCMNPARAFGPAVVANHWNFHWIYWLGPLLAGLLVGLLIRCFIGDGKTRLILKAR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 343
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.