missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin 7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577712
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Reconstitute using 50 uL deionized water.
Aquaporins/major intrinsic protein (MIP) are a family of water-selective membrane channels. Aquaporin 7 has greater sequence similarity with AQP3 and AQP9 and they may be a subfamily. Aquaporin 7 and AQP3 are at the same chromosomal location suggesting that 9p13 may be a site of an aquaporin cluster. Aquaporin 7 facilitates water, glycerol and urea transport. It may play an important role in sperm function.
Specifications
| Aquaporin 7 | |
| Polyclonal | |
| Unconjugated | |
| Aqp7 | |
| AQP7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11832, 29171 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| O54794, P56403 | |
| Aqp7 | |
| Peptide corresponding to amino acid residues 7-24 of rat AQP-7 | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction