missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aquaporin 6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578815
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: U87 Cell, COLO320 Cell, MCF-7 Cell. IHC: Human Placenta Tissue, Human Lung Cancer Tissue.
Aquaporin 6 forms a water-specific channel that participates in distinct physiological functions such as glomerular filtration, tubular endocytosis and acid-base metabolism.
Specifications
| Aquaporin 6 | |
| Polyclonal | |
| Unconjugated | |
| AQP6 | |
| AI790337; AQP2L; Aqp6; AQP-6; aquaporin 2-like, kidney specific; aquaporin 6; aquaporin 6, kidney specific; Aquaporin-2-like; Aquaporin-6; aquaporin-6, kidney specific; hKID; KID; Kidney-specific aquaporin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 363 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| Q13520 | |
| AQP6 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human Aquaporin 6 (264-282aa EPLKKESQPGSGAVEMESV). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction