missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aquaporin 5 Recombinant Rabbit Monoclonal Antibody (K01_1M98)

Product Code. 30828959
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828959 50 μL
30825411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828959 Supplier Invitrogen™ Supplier No. MA564697
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Aquaporin 5 Recombinant Monoclonal Antibody for Western Blot

Many isoforms of aquaporin have been identified in mammals, designated AQP0 through AQP10. Aquaporins are widely distributed and it is not uncommon for more than one type of AQP to be present in the same cell. Although most aquaporins are only permeable to water, AQP3, AQP7, AQP9 and one of the two AQP10 transcripts are also permeable to urea and glycerol. AQP2 is the only water channel that is activated by vasopressin to enhance water reabsorption in the kidney collecting duct. Aquaporins are involved in renal water absorption, generation of pulmonary secretions, lacrimation, and the secretion and reabsorption of cerebrospinal fluid and aqueous humor. In the lung, AQP5 is responsible for the majority of water transport across the apical membrane of type I alveolar epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 5
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1M98
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene AQP5
Gene Accession No. P55064
Gene Alias AQP 5; Aqp5; AQP-5; aquaporin 5; Aquaporin5; aquaporin-5; PPKB
Gene Symbols AQP5
Host Species Rabbit
Immunogen A synthetic peptide of human Aquaporin 5.
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.