Learn More
Aquaporin 4 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL594-16473-100UL
Description
The Aquaporin 4 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Aquaporin 4. This antibody recognizes human, mouse, rat antigen. The Aquaporin 4 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| Aquaporin 4 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |
| Immunofluorescence | |
| CoraLite 594 | |
| 100 μL | |
| 361 | |
| IgG |