Learn More
Aquaporin-4 Antibody (CL0178) [Allophycocyanin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52872APC
Product Code. 30295539
  • £558.00 / 0.10mL
Description
Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aquaporin-4
Monoclonal
APC
aquaporin 4, MGC22454
This antibody was developed using a recombinant protein derived from P55087, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
361
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0178
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified