missing translation for 'onlineSavingsMsg'
Learn More

Aquaporin-4 Antibody (AQP4/3324), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18364342
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18364342 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18364342 Supplier Bio-Techne Supplier No. NBP314104AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin-4
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AQP4/3324
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias aquaporin 4, MGC22454
Host Species Mouse
Immunogen Human recombinant Aquaporin-4 protein fragment (around aa254-323) (exact sequence is proprietary) (Uniprot: P55087)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 361
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.