Learn More
Aquaporin-4 Antibody (AQP4/3323), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-44612-100ug
 View more versions of this product
Product Code. 30293750
  • £368.00 / 100µg
Description
Aquaporin-4 Monoclonal antibody specifically detects Aquaporin-4 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aquaporin-4
Monoclonal
0.2 mg/ml
10 mM PBS with 0.05% BSA
Mouse
Protein G purified
RUO
Primary
Human
Purified
Flow Cytometry, Immunohistochemistry, Western Blot
AQP4/3323
Unconjugated
aquaporin 4, MGC22454
Human recombinant Aquaporin-4 protein fragment (around aa200-323) (Uniprot: P55087). Exact sequence is proprietary.
100 μg
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
361
Store at 4⪚C.
IgG1 κ