Learn More
Invitrogen™ Aquaporin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578811
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, rat kidney tissue, mouse kidney tissue. IHC: mouse kidney tissue, rat kidney tissue, human bladder cancer tissue. ICC/IF: A431 cell. Flow: RT4 cell.
AQP3 is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 3 is localized at the basal lateral membranes of collecting duct cells in the kidney. In addition to its water channel function, aquaporin 3 has been found to facilitate the transport of nonionic small solutes such as urea and glycerol, but to a smaller degree. It has been suggested that water channels can be functionally heterogeneous and possess water and solute permeation mechanisms.
Specifications
| Aquaporin 3 | |
| Polyclonal | |
| Unconjugated | |
| AQP3 | |
| 31.4 kDa water channel protein; AQP 3; AQP-2; AQP3; AQP-3; Aquaglyceroporin-3; aquaporin 3; aquaporin 3 (GIL blood group); aquaporin 3 (Gill blood group); Aquaporin3; aquaporin-3; GIL; Gill blood group | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11828, 360, 65133 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| Antibody with 0.2mg Na2PO4, 0.9mg NaCl, 4mg trehalose and no preservative | |
| P47862, Q8R2N1, Q92482 | |
| AQP3 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human Aquaporin 3 (278-292aa EENVKLAHVKHKEQI). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.