missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
£180.00
Specifications
| Antigen | Aquaporin-3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
Description
Aquaporin-3 Polyclonal specifically detects Aquaporin-3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Aquaporin-3 | |
| Western Blot 1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:400, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Rabbit | |
| Membrane Trafficking and Chaperones | |
| PBS and 50% Glycerol with 0.09% Sodium Azide | |
| AQP-3, Aquaglyceroporin-3, aquaporin 3, aquaporin 3 (GIL blood group), aquaporin 3 (Gill blood group), aquaporin-3, GIL, Gill blood group | |
| AQP3 | |
| IgG | |
| Affinity Purified | |
| Detects approx 31.5kDa. May detect larger glycosylated bands approx 35-50kDa. |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| P47862 | |
| 360 | |
| Produced against the C-terminal peptide (Sequence N-CHLEQPPPSTEAENVKLAHMKHKEQI) of rat aquaporin 3 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| 32 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title