Learn More
Invitrogen™ Aquaporin 2 Recombinant Rabbit Monoclonal Antibody (24GB6835)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556968
 View more versions of this product
Product Code. 30274879
  • £126.00 / 20µL
Description
Aquaporin 2 Recombinant Monoclonal Antibody for Western Blot
Aquaporin 2 (AQP2) is a hormonally regulated water channel located in the renal collecting duct. Mutations in the AQP2 gene cause hereditary nephrogenic diabetes insipidus in humans. A vasopressin induced cAMP increase results in the phosphorylation of AQP2 at serine-256 and its translocation from the intracellular vesicles to the apical membrane of principal cells. Recently, serine-261 has been identified as a novel phosphorylation site on AQP2 and levels of phosphorylated S261 have been shown to decrease with vasopressin treatment suggesting its involvement in vasopressin-dependent AQP2 trafficking.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aquaporin 2
Recombinant Monoclonal
Unconjugated
Aqp2
ADH water channel; AQP 2; AQP CD; Aqp2; AQP-2; AQPCD; AQP-CD; aquaporin 2; aquaporin 2 (collecting duct); Aquaporin CD; Aquaporin2; aquaporin-2; Aquaporin-CD; Aquaporine 2; collecting duct water channel protein; cph; jpk; MGC34501; Water channel protein for renal collecting duct; water-channel aquaporin 2; WCH CD; WCHCD; WCH-CD
Rabbit
Affinity chromatography
RUO
359
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB6835
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P41181
Aqp2
A synthesized peptide derived from human Aquaporin 2 (aa 191-202).
20 μL