missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aquaporin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578808
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: MCF-7 Cell, SW620 Cell, HT1080 Cell. IHC: Rat Kidney Tissue IHC-F: Rat Kidney Tissue.
Aquaporin 2 (AQP2) is a hormonally regulated water channel located in the renal collecting duct. Mutations in the AQP2 gene cause hereditary nephrogenic diabetes insipidus in humans. A vasopressin induced cAMP increase results in the phosphorylation of AQP2 at serine-256 and its translocation from the intracellular vesicles to the apical membrane of principal cells. Recently, serine-261 has been identified as a novel phosphorylation site on AQP2 and levels of phosphorylated S261 have been shown to decrease with vasopressin treatment suggesting its involvement in vasopressin-dependent AQP2 trafficking.
Specifications
| Aquaporin 2 | |
| Polyclonal | |
| Unconjugated | |
| Aqp2 | |
| ADH water channel; AQP 2; AQP CD; Aqp2; AQP-2; AQPCD; AQP-CD; aquaporin 2; aquaporin 2 (collecting duct); Aquaporin CD; Aquaporin2; aquaporin-2; Aquaporin-CD; Aquaporine 2; collecting duct water channel protein; cph; jpk; MGC34501; Water channel protein for renal collecting duct; water-channel aquaporin 2; WCH CD; WCHCD; WCH-CD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11827, 25386, 359 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| P34080, P41181, P56402 | |
| Aqp2 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human Aquaporin 2 (245-260aa DWEEREVRRRQSVELH). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction