missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aquaporin 10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109588
This item is not returnable.
View return policy
Description
Immunogen sequence: CGIPLNPARD LGPRLFTYVA GWGPEVFSAG NGWWWVPVVA PLVGATVGTA TYQLLVALHH PEGPEPAQDL VSAQHKASEL ETPASAQMLE CKL.
AQP10 is a member of the aquaglyceroporin family of integral membrane proteins. Members of this family function as water-permeable channels in the epithelia of organs that absorb and excrete water. AQP10 was shown to function as a water-selective channel, and could also permeate neutral solutes such as glycerol and urea. This gene encodes a member of the aquaglyceroporin family of integral membrane proteins. Members of this family function as water-permeable channels in the epithelia of organs that absorb and excrete water. This protein was shown to function as a water-selective channel, and could also permeate neutral solutes such as glycerol and urea.
Specifications
| Aquaporin 10 | |
| Polyclonal | |
| Unconjugated | |
| AQP10 | |
| AQP10; AQP-10; AQPA_HUMAN; Aquaglyceroporin-10; aquaporin 10; aquaporin-10; Small intestine aquaporin | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 89872 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96PS8 | |
| AQP10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 209-301 of human AQP10 (NP_536354.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction