missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-0 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £470.00
Specifications
| Antigen | Aquaporin-0 |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18259263
|
Novus Biologicals
NBP2-58661 |
100 μL |
£470.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615348
|
Novus Biologicals
NBP2-58661-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aquaporin-0 Polyclonal specifically detects Aquaporin-0 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Aquaporin-0 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| aquaporin 0, lens fiber major intrinsic protein, major intrinsic protein of lens fiber, MP26 | |
| MIP | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 4284 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RLKSISERLSVLKGAKPDVSNGQPEVTGEPVELNTQAL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title