missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin-0 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92773-0.1ml
This item is not returnable.
View return policy
Description
Aquaporin-0 Polyclonal antibody specifically detects Aquaporin-0 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Aquaporin-0 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:200, Immunohistochemistry-Paraffin | |
| aquaporin 0, lens fiber major intrinsic protein, major intrinsic protein of lens fiber, MP26 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human MIP (NP_036196.1). LALATLVQSVGHISGAHVNPAVTFAFLVGSQMSLLRAFCYMAAQLLGAVAGAAVLYSVTPPAVRGNLALNTLHPAVSVGQATTVEIFLTLQFVLCIFATYD | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Endocrinology, Neuroscience, Signal Transduction | |
| 4284 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction