missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Solvent, for Karl Fischer Titration By Volumetry, Fisher Chemical

Solvent for two-component system; Pyridine free

Brand:  Fisher Chemical K/2100/17

Code : 24

Additional Details : CAS Number : 67-56-1 Weight : 2.50000kg Transport : UN number : 3286 Chem class : 1a Pack group : II

Quantity 2.5L
Packaging Amber glass bottle

 View more versions of this product

Product Code. 10264390

  • £157.33 / 2.50 L


Estimated Shipment
22-02-2018
Add to basket


Description

Description

  • Used in conjunction with Aqualine™ titrant for two component volumetric Karl Fischer analysis
Specifications

Chemical Identifiers

7446-09-5
Clear colorless
81734
Amber glass bottle
1992
> 1.0
Aqualine™ Solvent
Liquid
Karl Fischer Reagent
Pyridine free
Pass Test

Specifications

Aqualine™ Solvent
Clear colorless
Karl Fischer Reagent
Amber glass bottle
> 1.0
81734
Pyridine free
2.5L
7446-09-5
Pass Test
Liquid
1992
Videos
SDS Documentation