Learn More
Karl Fischer Aqualine™ Solvent, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Solvent for two-component system; Pyridine free
Brand: Fisher Chemical K/2100/15
80.26 GBP valid until 2024-03-30
Use promo code "21741" to get your promotional price.
Code : 24
Additional Details : CAS Number : 67-56-1 Weight : 1.44000kg Transport : UN number : 3286 Chem class : 3 Pack group : II
Description
Solvent for two-component system; Pyridine free
- Used in conjunction with Aqualine™ titrant for two component volumetric Karl Fischer analysis
Specifications
Colorless | |
Pyridine free | |
Karl Fischer Reagent | |
Amber glass bottle | |
Pass Test | |
81734 |
1 L | |
Aqualine™ Solvent | |
288-32-4,7446-09-5 | |
Liquid | |
> 1.0 | |
1992 |
Safety and Handling
- Aq Solvent, 4
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.