missing translation for 'onlineSavingsMsg'
Learn More

Aqualine™ Complete 2, Fisher Chemical™

Product Code. p-305664 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
Color:
Brown
Colorless
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10744761

Brand: Fisher Chemical K/1950/15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Single-component reagent; Pyridine free

  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 2.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free
TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Physical Form Liquid
Quantity 1 L
MDL Number 81734
Packaging Amber glass bottle
compliance-icons compliance-icons
Product Identifier
  • Aqualine Complete 2
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Iodine, 1-Imidazole,Diethylene glycol monoethyl ether
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.