Learn More
Macherey Nagel Aquadur™ Water Hardness Test Strips
For semi-quantitative determination of water hardness (CaC03)
Brand: Macherey Nagel 91239
This item is not returnable.
View return policy
Description
- Test papers allow the semi-qualitative determination of water hardness
- Ranged test results
Specifications
| CaCO3 | |
| 100 x 6 x 95 mm Strips | |
| <54, >72, >151.2, >252 ppm | |
| Test strips, semi-quantitative, water hardness |