missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AQR Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24342-1-AP-150UL
This item is not returnable.
View return policy
Description
Intron-binding spliceosomal protein required to link pre-mRNA splicing and snoRNP (small nucleolar ribonucleoprotein) biogenesis. Plays a key role in position-dependent assembly of intron-encoded box C/D small snoRNP, splicing being required for snoRNP assembly. May act by helping the folding of the snoRNA sequence. Binds to intron of pre-mRNAs in a sequence-independent manner, contacting the region between snoRNA and the branchpoint of introns (40 nucleotides upstream of the branchpoint) during the late stages of splicing.Specifications
| AQR | |
| Polyclonal | |
| Unconjugated | |
| AQR | |
| AQR, aquarius homolog (mouse), fSAP164, IBP160, KIAA0560 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9716 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60306 | |
| AQR | |
| AQR Fusion Protein Ag17810 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction