missing translation for 'onlineSavingsMsg'
Learn More

Apyramide, MedChemExpress

Product Code. 16452328
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
20 mg
Unit Size:
10mg
20mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452328 5 mg 5mg
16420656 20 mg 20mg
16400139 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16452328 Supplier Medchem Express Supplier No. HYU00046
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Apyramide is an anti-inflammatory agent (NSAID) and behaves as a prodrug of indomethacin (HY-14397). Indomethacin is a potent, blood-brain permeable and nonselective inhibitor of COX1 and COX2.

Apyramide is an anti-inflammatory agent (NSAID) and behaves as a prodrug of indomethacin (HY-14397). Indomethacin is a potent, blood-brain permeable and nonselective inhibitor of COX1 and COX2.

Specifications

Chemical Name or Material Apyramide
CAS 68483-33-0
Color White
Purity Grade Notes Research
Molecular Formula C27H23ClN2O5
Quantity 5 mg
SMILES O=C(OC1=CC=C(NC(C)=O)C=C1)CC2=C(C)N(C(C3=CC=C(Cl)C=C3)=O)C4=C2C=C(OC)C=C4
Molecular Weight (g/mol) 490.93
Formula Weight 490.93
Percent Purity 99.06%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.