missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ APTS (8-Aminopyrene-1,3,6-Trisulfonic Acid, Trisodium Salt)
Click to view available options
Quantity:
10 mg
Packaging:
1 Each
Unit Size:
10mg
Description
The aromatic amine of APTS (8-aminopyrene-1,3,6-trisulfonic acid, trisodium salt) can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new biotinylated probes.
Specifications
Specifications
| Melting Point | >250°C |
| Color | Yellow |
| Packaging | 1 Each |
| Quantity | 10 mg |
| Physical Form | Solid |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction