missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford APS Tablets 100 Tab
Description
Used as a catalyst for acrylamide gel polymerization.
Used as a catalyst for acrylamide gel polymerization.
Specifications
Specifications
| Chemical Name or Material | APS Tablets, 100 Tablets |
| CAS | 7727-54-0 |
| Quantity | 100 Tablets |
| Molecular Formula | H8N2O8S2 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?