missing translation for 'onlineSavingsMsg'
Learn More

APS-2-79, MedChemExpress

Product Code. 16453108
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453108

Brand: Medchem Express HY100627

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress APS-2-79 is a KSR-dependent MEK antagonist. APS-2-79 inhibits ATPbiotin binding to KSR2 within the KSR2-MEK1 complexe with an IC50 of 120 nM. APS-2-79 makes the stabilization of the KSR inactive state antagonizes oncogenic Ras-MAPK signaling.

APS-2-79 is a KSR-dependent MEK antagonist. APS-2-79 inhibits ATPbiotin binding to KSR2 within the KSR2-MEK1 complexe with an IC50 of 120 nM. APS-2-79 makes the stabilization of the KSR inactive state antagonizes oncogenic Ras-MAPK signaling.

Specifications

Chemical Name or Material APS-2-79
CAS 2002381-25-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H21N3O3
Quantity 10 mg
Solubility Information DMSO : 20 mg/mL (51.62 mM; Need ultrasonic)
SMILES CC1=CC(OC2=CC=CC=C2)=CC=C1NC3=NC=NC4=CC(OC)=C(OC)C=C43
Molecular Weight (g/mol) 387.43
Formula Weight 387.43
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.