missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ APRT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578804
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, human A549 whole cell, human K562 whole cell. ICC/IF: MCF-7 cell. Flow: A549 cell.
Adenine phosphoribosyltransferase belongs to the purine/pyrimidine phosphoribosyltransferase family. A conserved feature of this gene is the distribution of CpG dinucleotides. This enzyme catalyzes the formation of AMP and inorganic pyrophosphate from adenine and 5-phosphoribosyl-1-pyrophosphate (PRPP). It also produces adenine as a by-product of the polyamine biosynthesis pathway. A homozygous deficiency in this enzyme causes 2,8-dihydroxyadenine urolithiasis. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| APRT | |
| Polyclonal | |
| Unconjugated | |
| Aprt | |
| adenine phosphoribosyl transferase; adenine phosphoribosyltransferase; AMP; AMP diphosphorylase; AMP pyrophosphorylase; APRT; APRTD; C85684; DKFZp686D13177; MGC125856; MGC125857; MGC129961; transphosphoribosidase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 353 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Western Blot, Immunocytochemistry, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P07741 | |
| Aprt | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human APRT (5-49aa ELQLVEQRIRSFPDFPTPGVVFRDISPVLKDPASFRAAIGLLARH). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction