Learn More
Invitrogen™ Aprotinin Monoclonal Antibody (21)

Mouse Monoclonal Antibody
Brand: Invitrogen™ ABS0372102
Description
ABS 037-21-02 detects aprotinin in bovine samples. ABS 037-21-02 has successfully been used in ELISA applications. PBS based buffers are unsuitalbe for this assay and 0.2 M glycine sodium buffer, pH 8.8, is recommended for washing and dilution buffers. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
Aprotinin, also called bovine pancreatic trypsin inhibitor, is a single chain globular Kunitz-type serine proteinase inhibitor found in bovine pancreas, parotid gland and lung. It forms stable, enzymatically inactive complexes with trypsin, chymotrypsin, plasmin and kallikrein. Aprotinin has been used therapeutically as an antifibrinolytic agent under the name Trasylol (Bayer) to reduce blood loss during cardiopulmonary bypass surgery but this use has been associated with an increased frequency of acute renal injury.
Specifications
| Aprotinin | |
| Monoclonal | |
| 0.94 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| 0 | |
| PTI | |
| Aprotinin from bovine lung. | |
| 400 μL | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 21 | |
| Unconjugated | |
| PTI | |
| aprotinin; basic protease inhibitor; BPI; BPTI; pancreatic trypsin inhibitor; PTI; trypsin ihibitor; trypsin ihibitor precursor | |
| Mouse | |
| Protein A | |
| RUO | |
| 404172 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.