Learn More
Tocris Bioscience™ Aprotinin
GREENER_CHOICE
Product Code. 18775688
Click to view available options
:
10mg
Product Code. 18775688
Brand: Tocris Bioscience™ 4139/10
Competitive serine protease inhibitor
Aprotinin is a competitive serine protease inhibitor. Reversibly binds to and blocks the enzymatic active site. Inhibits a range of serine proteases including trypsin, chymotrypsin, kallikrein and plasmin. Inhibits cytopathogenic effect of SARS-CoV-2 and double-stranded RNA formation in SARS-CoV-2-infected cells.
This product is from Bovine Lung.
TRUSTED_SUSTAINABILITY
Specifications
Molecular Weight (g/mol) 6511.48
Product Type Aprotinin
Quantity 10 mg