missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Aprotinin
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
Description
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes.
Competitive serine protease inhibitor which forms stable complexes with and blocks the active sites of enzymes. The binding is reversible, and most aprotinin-protease complexes dissociate at a pH>10 or <3.2. Target enzumes include trypsin, chymatrypsin, kalikrein and plasmin. Activity: >4500 KIU/mg
Specifications
Specifications
| CAS | 9087-70-1 |
| Molecular Formula | C284H432N84O79S7 |
| Quantity | 50 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction