missing translation for 'onlineSavingsMsg'
Learn More

Aprocitentan, MedChemExpress

Product Code. p-200100598
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16499437

Brand: Medchem Express HY15895

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Aprocitentan (ACT-132577) is the major and pharmacologically active metabolite of Macitentan. Aprocitentan is dual ETA/ETB antagonist with IC50s of 3.4 nM and 987 nM, and pA2 valus of 6.7 and 5.5, respectively.

Aprocitentan (ACT-132577) is the major and pharmacologically active metabolite of Macitentan. Aprocitentan is dual ETA/ETB antagonist with IC50s of 3.4 nM and 987 nM, and pA2 valus of 6.7 and 5.5, respectively.

Specifications

Chemical Name or Material Aprocitentan
CAS 1103522-45-7
Color White
Purity Grade Notes Research
Molecular Formula C16H14Br2N6O4S
Quantity 5 mg
Synonym ACT-132577
Solubility Information DMSO : ≥ 46 mg/mL (84.22 mM)
SMILES O=S(N)(NC1=NC=NC(OCCOC2=NC=C(Br)C=N2)=C1C3=CC=C(Br)C=C3)=O
Molecular Weight (g/mol) 546.19
Formula Weight 546.19
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.