missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APRIL/TNFSF13 Antibody (SR1578), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22069-50ul
This item is not returnable.
View return policy
Description
APRIL/TNFSF13 Monoclonal antibody specifically detects APRIL/TNFSF13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| APRIL/TNFSF13 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1578 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2 | |
| A synthesized peptide derived from human APRIL/TNFSF13 (Uniprot #: O75888) | |
| 50 μg | |
| Apoptosis, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology | |
| 8741 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction