missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ APRIL/TNFSF13 Antibody (sibeprenlimab) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
£162.00 - £695.00
Specifications
| Antigen | APRIL/TNFSF13 |
|---|---|
| Clone | sibeprenlimab |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30047288
|
Novus Biologicals™
NBP3-28614-100ug |
100 μg |
£247.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30045697
|
Novus Biologicals™
NBP3-28614-1mg |
1 mg |
£695.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30047186
|
Novus Biologicals™
NBP3-28614-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
APRIL/TNFSF13 Monoclonal antibody specifically detects APRIL/TNFSF13 in Human samples. It is validated for Flow Cytometry,ELISA,Functional AssaySpecifications
| APRIL/TNFSF13 | |
| Flow Cytometry, ELISA, Functional Assay | |
| Unconjugated | |
| Human | |
| Apoptosis, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2 | |
| TNFSF13/ APRIL/ CD256 | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. |
| sibeprenlimab | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| O75888 | |
| 8741 | |
| IgG2 | |
| Protein A purified | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title