missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ APRIL/TNFSF13 Antibody (BION-1301) - Humanized, IgG2SA, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28382-1mg
This item is not returnable.
View return policy
Description
APRIL/TNFSF13 Monoclonal antibody specifically detects APRIL/TNFSF13 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| APRIL/TNFSF13 | |
| Monoclonal | |
| Unconjugated | |
| O75888 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG2 |
| Flow Cytometry, ELISA, Functional Assay | |
| BION-1301 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2 | |
| TNFSF13/ APRIL/ CD256 | |
| 1 mg | |
| Apoptosis, Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research, Immunology | |
| 8741 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction