Learn More
PeproTech APRIL Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Description
AA Sequence of recombinant protein: AVLTQKQKKQ HSVLHLVPIN ATSKDDSDVT EVMWQPALRR GRGLQAQGYG VRIQDAGVYL LYSQVLFQDV TFTMGQVVSR EGQGRQETLF RCIRSMPSHP DRAYNSCYSA GVFHLHQGDI LSVIIPRARA KLNLSPHGTF LGFVKL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human APRIL. Anti-Human APRIL-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hAPRIL by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human APRIL (500-P192) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hAPRIL. Western Blot: To detect Human APRIL by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human APRIL is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P192BT-1MG will be provided as 2 x 500 μg
Specifications
Specifications
| Antigen | APRIL |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1-1.0 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with no preservative |
| Gene | TNFSF13 |
| Gene Accession No. | O75888 |
| Gene Alias | 2310026N09Rik; A proliferation-inducing ligand; APRIL; CD256; TALL2; TALL-2; TNF- and APOL-related leukocyte expressed ligand 2; TNF-related death ligand 1; Tnfsf13; TNLG7B; Trdl1; TRDL-1; tumor necrosis factor (ligand) superfamily, member 13; tumor necrosis factor ligand 7B; tumor necrosis factor ligand superfamily member 13; tumor necrosis factor superfamily member 13; tumor necrosis factor-like protein ZTNF2; tumor necrosis factor-related death ligand-1; UNQ383/PRO715; ZTNF2 |
| Gene Symbols | TNFSF13 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.