missing translation for 'onlineSavingsMsg'
Learn More

Aprepitant, MedChemExpress

Product Code. 16483058
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483058 10 mg 10mg
16439626 50 mg 50mg
16435797 5 mg 5mg
16410916 200 mg 200mg
16404366 100 mg 100mg
16401526 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16483058 Supplier Medchem Express Supplier No. HY10052
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Aprepitant (MK-0869) is a selective and high-affinity neurokinin 1 receptor antagonist with a Kd of 86 pM.

Aprepitant (MK-0869) is a selective and high-affinity neurokinin 1 receptor antagonist with a Kd of 86 pM.

Specifications

Chemical Name or Material Aprepitant
CAS 170729-80-3
Color White
Purity Grade Notes Research
Molecular Formula C23H21F7N4O3
Quantity 10 mg
Synonym MK-0869 MK-869 L-754030
Solubility Information DMSO : ≥ 100 mg/mL (187.12 mM)
SMILES O=C1NC(CN2[C@H]([C@@H](O[C@@H](C3=CC(C(F)(F)F)=CC(C(F)(F)F)=C3)C)OCC2)C4=CC=C(F)C=C4)=NN1
Molecular Weight (g/mol) 534.43
Formula Weight 534.43
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.