missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Aprepitant
GREENER_CHOICE

Product Code. 18717276
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18717276

Brand: Tocris Bioscience™ 6486/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent long-acting hNK-1 antagonist

Aprepitant is a potent long-acting hNK-1 antagonist, (IC50 = 0.09 nM and Kd = 19 pM). Inhibits induced foot-tapping in gerbils. CNS penetrant and orally bioavailable. Antiemetic.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[[(2R,3S)-2-[(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl)-4-morpholinyl]methyl]-1,2-dihydro-3H-1,2,4-triazol-3-one
CAS 170729-80-3
Quantity 50 mg
Molecular Formula C23H21F7N4O3
Purity 99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.