missing translation for 'onlineSavingsMsg'
Learn More

Apremilast, TRC

Product Code. 30008272
Change view
Click to view available options
Quantity:
5 mg
100 mg
2.5 g
Unit Size:
100mg
2.5g
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008272 2.5 g 2.5g
30007307 100 mg 100mg
30004852 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008272 Supplier Toronto Research Chemicals Supplier No. TRCA7297002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608141-41-9
Formula Weight 460.1304
InChI Formula InChI=1S/C22H24N2O7S/c1-5-31-19-11-14(9-10-18(19)30-3)17(12-32(4,28)29)24-21(26)15-7-6-8-16(23-13(2)25)20(15)22(24)27/h6-11,17H,5,12H2,1-4H3,(H,23,25)/t17-/m1/s1
IUPAC Name N-[2-[(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-methylsulfonylethyl]-1,3-dioxoisoindol-4-yl]acetamide
Molecular Formula C22 H24 N2 O7 S
Molecular Weight (g/mol) 460.5
Percent Purity >95%
SMILES CCOc1cc(ccc1OC)[C@@H](CS(=O)(=O)C)N2C(=O)c3cccc(NC(=O)C)c3C2=O
Synonym Acetamide, N-[2-[(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]-, N-[2-[(1S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]-2,3-dihydro-1,3-dioxo-1H-isoindol-4-yl]acetamide, (S)-2-[1-(3-Ethoxy-4-methoxyphenyl)-2-methylsulfonylethyl]-4-acetylaminoisoindoline-1,3-dione, (S)-N-[2-[1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl]ethyl]-1,3-dioxoisoindolin-4-yl)acetamide, Apremilast, CC 10004, Otezla
Chemical Name or Material Apremilast
Grade HPLC
Show More Show Less

Specifications

Quantity 2.5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.