missing translation for 'onlineSavingsMsg'
Learn More

Apremilast open ring 3-acetamide, TRC

Product Code. 30340265
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30340265

Brand: Toronto Research Chemicals TRCA72971525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Acetylamino)-2-((((1S)-1-(3-ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl)amino)carbonyl)benzoic Acid (Apremilast Benzoic Acid)
CAS 2096492-41-8
Molecular Formula C22H26N2O8S
Quantity 25 mg
Synonym 3-(Acetylamino)-2-[[[(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]amino]carbonyl]benzoic Acid (Apremilast Benzoic Acid), Apremilast Hydrolysis Impurity-2
InChI Formula InChI=1S/C22H26N2O8S/c1-5-32-19-11-14(9-10-18(19)31-3)17(12-33(4,29)30)24-21(26)20-15(22(27)28)7-6-8-16(20)23-13(2)25/h6-11,17H,5,12H2,1-4H3,(H,23,25)(H,24,26)(H,27,28)/t17-/m1/s1
SMILES CCOc1cc(ccc1OC)[C@@H](CS(=O)(=O)C)NC(=O)c2c(NC(=O)C)cccc2C(=O)O
IUPAC Name 3-acetamido-2-[[(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-methylsulfonylethyl]carbamoyl]benzoic acid
Molecular Weight (g/mol) 478.52
Formula Weight 478.141
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.